Ácido 2-fluoro-3-metoxifenilborónico (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-Fluoro-3-methoxybenzeneboronic acid, (2-Fluoro-3-methoxyphenyl)boronic acid
F0940-1G
F0940-5G
TCIAF0940-1GEA
16
EUR
TCIAF0940-1G
TCIAF0940-5G
Ácido 2-fluoro-3-metoxifenilborónico (contains varying amounts of Anhydride)
Ácido 2-fluoro-3-metoxifenilborónico
Formula:
C₇H₈BFO₃ MW: 169,95 g/mol |
Número MDL:
MFCD02094728 Núm. CAS: 352303-67-4 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 112.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...