3-Fluoro-4'-n-propylbiphenyl-4-boronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
3-Fluoro-4'-propyl-4-biphenylboronic acid
F0942-1G
F0942-5G
TCIAF0942-1GEA
40
EUR
TCIAF0942-1G
TCIAF0942-5G
3-Fluoro-4'-n-propylbiphenyl-4-boronic acid (contains varying amounts of Anhydride)
3-Fluoro-4'-n-propylbiphenyl-4-boronic acid
Formula:
C₁₅H₁₆BFO₂ MW: 258,099 g/mol |
Número MDL:
MFCD10687198 Núm. CAS: 909709-42-8 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 97.0 to 108.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...