Fenofibric acid ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-(4-(4-Chlorobenzoyl)phenoxy)-2-methylpropanoic acid
F1011-1G
F1011-5G
TCIAF1011-1GEA
33
EUR
TCIAF1011-1G
TCIAF1011-5G
Fenofibric acid ≥98.0% (by GC, titration analysis)
Fenofibric acid
Formula:
C₁₇H₁₅ClO₄ MW: 318,75 g/mol |
Número MDL:
MFCD00792461 Núm. CAS: 42017-89-0 EINECS: 255-626-9 UN: 3077 ADR: 9,III |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 179.0 to 183.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...