Fmoc-L-Tyr(Me)-OH ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
Fmoc-Tyr(Me)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-methyl-L-tyrosine, N-Fmoc-4-methoxy-L-phenylalanine, N-Fmoc-O-methyl-L-tyrosine, Fmoc-Phe(4-OMe)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-methoxy-L-phenylalanine
F1038-1G
F1038-5G
TCIAF1038-1GEA
48
EUR
TCIAF1038-1G
TCIAF1038-5G
Fmoc-L-Tyr(Me)-OH ≥98.0% (by HPLC, titration analysis)
Fmoc-L-Tyr(Me)-OH
Formula:
C₂₅H₂₃NO₅ MW: 417.47 g/mol |
Número MDL:
MFCD00153368 Núm. CAS: 77128-72-4 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 159.0 to 163.0 deg-C |
Specific rotation [a]20/D | -26.0 to -30.0 deg (C=1,DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...