5-(4-Fluorophenyl)-2-thiophenecarboxylic acid ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
F1073-1G
F1073-5G
TCIAF1073-1GEA
40
EUR
TCIAF1073-1G
TCIAF1073-5G
5-(4-Fluorophenyl)-2-thiophenecarboxylic acid ≥98.0% (by GC, titration analysis)
5-(4-Fluorophenyl)-2-thiophenecarboxylic acid
Formula:
C₁₁H₇FO₂S |
Número MDL:
MFCD01316570 Núm. CAS: 115933-30-7 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 248.0 to 252.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...