4-Fluoro-1-naphthaleneboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-Fluoronaphtalene-1-boronic acid, 4-Fluoronaphthalene-1-boronic acid
F1080-200MG
F1080-1G
TCIAF1080-200MGEA
18
EUR
TCIAF1080-200MG
TCIAF1080-1G
4-Fluoro-1-naphthaleneboronic acid (contains varying amounts of Anhydride)
4-Fluoro-1-naphthaleneboronic acid
Formula:
C₁₀H₇BBrFO₂ MW: 189,98 g/mol |
Número MDL:
MFCD04039221 Núm. CAS: 182344-25-8 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | 97.0 to 111.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...