N-(4-Fluorobenzyl)-3-nitro-p-phenylenediamine ≥97.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
N1-(4-Fluorobenzyl)-3-nitrobenzene-1,4-diamine, N1-(4-Fluorobenzyl)-3-nitro-1,4-phenylenediamine
F1082-1G
F1082-5G
TCIAF1082-1GEA
31
EUR
TCIAF1082-1G
TCIAF1082-5G
N-(4-Fluorobenzyl)-3-nitro-p-phenylenediamine ≥97.0% (by HPLC, titration analysis)
N-(4-Fluorobenzyl)-3-nitro-p-phenylenediamine
Formula:
C₁₃H₁₂FN₃O₂ MW: 261,256 g/mol |
Número MDL:
MFCD12828054 Núm. CAS: 150812-21-8 |
Resultados test especificaciones
Appearance | Orange to Red powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 110.0 to 114.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...