Fmoc-D-Thr(tBu)-OH ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-tert-butyl-D-threonine, Fmoc-D-Thr(tBu)-OH, (2R,3S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(tert-butoxy)butanoic acid, N-Fmoc-O-tert-butyl-D-threonine
F1128-1G
F1128-5G
TCIAF1128-1GEA
24
EUR
TCIAF1128-1G
TCIAF1128-5G
Fmoc-D-Thr(tBu)-OH ≥98.0% (by HPLC, titration analysis)
Fmoc-D-Thr(tBu)-OH
Formula:
C₂₃H₂₇NO₅ MW: 397,471 g/mol |
Número MDL:
MFCD00080292 Núm. CAS: 138797-71-4 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 131.0 to 135.0 deg-C |
Specific rotation [a]20/D | -14.0 to -17.0 deg (C=1, EtOAc) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...