(3-Fluoro-4-hydroxyphenyl)boronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: 3-Fluoro-4-hydroxyphenylboronic acid, 3-Fluoro-4-hydroxybenzeneboronic acid

F1132-1G F1132-5G
TCIAF1132-1GEA 104 EUR
TCIAF1132-1G TCIAF1132-5G
(3-Fluoro-4-hydroxyphenyl)boronic acid (contains varying amounts of Anhydride)
(3-Fluoro-4-hydroxyphenyl)boronic acid
Formula: C₅H₅BFNO₂
MW: 155,92 g/mol
Número MDL: MFCD06659838
Núm. CAS: 182344-14-5

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 98.0 to 113.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR