Fmoc-N-methyl-L-norvaline ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: Fmoc-N-Me-Nva-OH, N-Fmoc-N-methyl-L-norvaline, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-norvaline, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)pentanoic acid

F1166-1G F1166-5G
TCIAF1166-1GEA 44 EUR
TCIAF1166-1G TCIAF1166-5G
Fmoc-N-methyl-L-norvaline ≥98.0% (by HPLC, titration analysis)
Fmoc-N-methyl-L-norvaline
Formula: C₂₁H₂₃NO₄
MW: 353,418 g/mol
Número MDL: MFCD01861326
Núm. CAS: 252049-05-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eeec953c61fe3186a4b0b50621989ea1e987ec50&vid=eeec953c61fe3186a4b0b50621989ea1e987ec50&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 130.0 to 135.0 deg-C
Specific rotation [a]20/D -24.0 to -28.0 deg (C=1, DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR