Formic acid-triethylamine 5:2 addition compound ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

Sinónimos: Formic acid triethylamine complex 5:2, Formic acid - triethylamine (5:2) Azeotrope, TEAF

F1202-25ML F1202-100ML
TCIAF1202-25MLEA 54 EUR
TCIAF1202-25ML TCIAF1202-100ML
Formic acid-triethylamine 5:2 addition compound ≥98.0% (by titrimetric analysis)
Formic acid-triethylamine 5:2 addition compound
Formula: C₁₇H₄₀N₂O₁₀
MW: 432,51 g/mol
Número MDL: MFCD00040600
Núm. CAS: 15077-13-1
UN: 3265
ADR: 8,II

Order Now

Resultados test especificaciones

Appearance Colorless to Almost colorless clear liquid
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR