Fomesafen ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
5-[2-Chloro-4-(trifluoromethyl)phenoxy]-N-(methylsulphonyl)-2-nitrobenzamide
F1236-1G
F1236-25G
TCIAF1236-1GEA
31
EUR
TCIAF1236-1G
TCIAF1236-25G
Fomesafen ≥98.0% (by HPLC, titration analysis)
Fomesafen
Formula:
C₁₅H₁₀ClF₃N₂O₆S MW: 438,76 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD01632756 Núm. CAS: 72178-02-0 EINECS: 276-439-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 220.0 to 224.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...