4-Fluorobenzylamine hydrochloride ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 4-Fluorobenzylammonium chloride, (4-Fluorophenyl)methanamine hydrochloride

F1255-1G F1255-5G
TCIAF1255-1GEA 28 EUR
TCIAF1255-1G TCIAF1255-5G
4-Fluorobenzylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
4-Fluorobenzylamine hydrochloride
Formula: C₇H₈FN·HCl
MW: 161,60 g/mol
Número MDL: MFCD00012861
Núm. CAS: 659-41-6
EINECS: 211-534-0

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR