4-Fluoro-2-formylphenylboronic acid ≥98.0% (by HPLC, titration analysis) (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
F1260-1G
F1260-5G
TCIAF1260-1GEA
64
EUR
TCIAF1260-1G
TCIAF1260-5G
4-Fluoro-2-formylphenylboronic acid ≥98.0% (by HPLC, titration analysis) (contains varying amounts of Anhydride)
4-Fluoro-2-formylphenylboronic acid
Formula:
C₇H₆BFO₃ Pto. de fusión: 125°C |
Número MDL:
MFCD10697426 Núm. CAS: 825644-26-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | 98.0 to 112.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...