Formamidine hydroiodide ≥99.0% (by titrimetric analysis) for perovskite precursor
Proveedor: TCI
Sinónimos:
FAI, Formamidinium iodide
F1263-1G
F1263-25G
F1263-5G
TCIAF1263-1GEA
62
EUR
TCIAF1263-1G
TCIAF1263-25G
TCIAF1263-5G
Formamidine hydroiodide ≥99.0% (by titrimetric analysis) for perovskite precursor
Formamidine hydroiodide
Formula:
CH₄N₂·HI MW: 171.97 g/mol |
Número MDL:
MFCD28369273 Núm. CAS: 879643-71-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystaline |
Purity (Argentmetric Titration) | min. 99.0 % |
Iodine content | min. 99.0 % |
Purity (Trace metal basis) | min. 99.99 % |
Solubility in 2-Propanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...