Guanidine sulphamate ≥90.0% (by titrimetric analysis)

Proveedor: TCI
Atención

G0022-25G G0022-500G
TCIAG0022-25GEA 16 EUR
TCIAG0022-25G TCIAG0022-500G
Guanidine sulphamate ≥90.0% (by titrimetric analysis)
Guanidine sulphamate
Formula: CH₅N₃
MW: 156,16 g/mol
Número MDL: MFCD00060261
Núm. CAS: 50979-18-5
EINECS: 256-886-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR