Iron(II) gluconate hydrate ≥95.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: Iron(II) Gluconate Hydrate, Iron(II) D-gluconate dihydrate, Gluconic Acid Iron(II) Salt

G0038-25G G0038-500G
TCIAG0038-25GEA 17 EUR
TCIAG0038-25G TCIAG0038-500G
Iron(II) gluconate hydrate ≥95.0% (by titrimetric analysis)
Iron(II) gluconate hydrate
Formula: C₁₂H₂₄FeO₁₅
MW: 464,16 g/mol
Número MDL: MFCD00150872
Núm. CAS: 22830-45-1
Índice Merck: 14,04047

Order Now

Resultados test especificaciones

Appearance White to Amber to Dark green powder to crystal
Purity (Cerium redox Method) min. 95.0 %
Specific rotation [a]20/D +5.0 to +10.0 deg (C=1 H2O)
Solubility in Water very faint turbidity
Drying loss max. 13.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR