D-Glucosamic acid ≥98.0% (by titrimetric analysis)

Proveedor: TCI

G0042-1G G0042-5G
TCIAG0042-1GEA 110 EUR
TCIAG0042-1G TCIAG0042-5G
D-Glucosamic acid ≥98.0% (by titrimetric analysis)
D-Glucosamic acid
Formula: C₆H₁₃NO₆
Número MDL: MFCD00037764
Núm. CAS: 3646-68-2
EINECS: 222-879-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6b36de0516aa59660a2dc4e66fa4ce72a79f927&vid=a6b36de0516aa59660a2dc4e66fa4ce72a79f927&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystaline
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -13.5 to -15.5 deg (C=2.5, 1mol/L HCl)
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR