γ-L-Glutamyl-4-nitroanilide (GPNA) hydrate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

G0065-1G G0065-5G
TCIAG0065-1GEA 85 EUR
TCIAG0065-1G TCIAG0065-5G
γ-L-Glutamyl-4-nitroanilide (GPNA) hydrate ≥98.0% (by HPLC, titration analysis)
γ-L-Glutamyl-4-nitroanilide (GPNA) hydrate
Formula: C₁₁H₁₃N₃O₅·H₂O
Número MDL: MFCD00036218
Núm. CAS: 7300-59-6
EINECS: 230-748-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9451e276579edaf9decf7ce443bb2d63781a998e&vid=fff89ca895fcd0cac4802effd9d4301cf5cc661d&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +26.0 to +32.0 deg (C=2, 0.5mol/L HCl)
Water 6.0 to 8.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR