Monolaurin ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
2,3-Dihydroxypropyl dodecanoate, Glycerol alpha-Monolaurate
G0081-25G
G0081-5G
TCIAG0081-25GEA
100
EUR
TCIAG0081-25G
TCIAG0081-5G
Monolaurin ≥98.0% (por GC)
Monolaurin
Formula:
C₁₅H₃₀O₄ MW: 274,40 g/mol Pto. Ebullición: 186°C |
Número MDL:
MFCD00037815 Núm. CAS: 142-18-7 EINECS: 205-526-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 60.0 to 66.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...