Calcium glycerophosphate ≥95.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: Calcium 2,3-dihydroxypropyl phosphate, Glycerophosphoric Acid Calcium Salt

G0095-25G G0095-500G
TCIAG0095-25GEA 17 EUR
TCIAG0095-25G TCIAG0095-500G
Calcium glycerophosphate ≥95.0% (by titrimetric analysis)
Calcium glycerophosphate
Formula: C₃H₇CaO₆P
MW: 210,13 g/mol
Número MDL: MFCD00042643
Núm. CAS: 27214-00-2
EINECS: 248-328-5
Índice Merck: 14,01670

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 95.0 % (after drying)
Solubility in Water faint turbidity
Drying loss max. 13.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR