Glycyl-D-leucine ≥99.0% (by titrimetric analysis)

Proveedor: TCI

G0130-1G G0130-100MG
TCIAG0130-1GEA 68 EUR
TCIAG0130-1G TCIAG0130-100MG
Glycyl-D-leucine ≥99.0% (by titrimetric analysis)
Glycyl-D-leucine
Formula: C₈H₁₆N₂O₃
Pto. de fusión: 234°C
Número MDL: MFCD00066050
Núm. CAS: 688-13-1

Order Now

Resultados test especificaciones

Appearance White powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Specific rotation [a]20/D +33.0 to +37.0 deg (C=2, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR