Glycyl-DL-norvaline ≥97.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
H-Gly-DL-Nva-OH
G0134-1G
G0134-5G
TCIAG0134-1GEA
78
EUR
TCIAG0134-1G
TCIAG0134-5G
Glycyl-DL-norvaline ≥97.0% (by titrimetric analysis)
Glycyl-DL-norvaline
Formula:
C₇H₁₄N₂O₃ MW: 174,20 g/mol |
Número MDL:
MFCD00038186 Núm. CAS: 2189-27-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...