Glycyl-l-phenylalanine

Proveedor: TCI

G0136-1G G0136-100MG
TCIAG0136-1GEA 83 EUR
TCIAG0136-1G TCIAG0136-100MG
Glycyl-l-phenylalanine
Proteínas y Péptidos

  • Specific rotation [a]20/D: 41° (C=2, H₂O)
  • Solubility in water: almost transparency
  • EINECS: 222-027-9


Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-Gly-Phe-OH

Order Now

ESPECIFICACIONES

Nombre de proteína / péptido Glycyl-L-phenylalanine
Sinónimos de proteínas H-Gly-Phe-OH
Environmentally Preferable
Especies
Fuente
CAS No. 3321-03-7
MDL No. MFCD00065110
Pureza Min. 98,0% (HPLC, T)
Forma White to almost white powder to crystal
Molecular weight 222,24 g/mol
Molecular formula C₁₁H₁₄N₂O₃
Condiciones de almacenamiento Refrigerator

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR