Glycyl-l-tryptophan hydrate
Proveedor: TCI
G0144-1G
G0144-100MG
TCIAG0144-1GEA
34
EUR
TCIAG0144-1G
TCIAG0144-100MG
Glycyl-l-tryptophan hydrate
Proteínas y Péptidos
- Specific rotation [a]20/D: 27,0 to 30,0° (C=2, 5 mol/L HCl)
- Solubility in 5 mol/L HCl: almost transparency
- EINECS: 219-235-7
Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-Gly-Trp-OH
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...