Glycocyamine ≥97.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Glycocyamin, N-Amidinoglycine, 2-Guanidinoacetic acid, Guanidinoacetic Acid, Guanidoacetic Acid
G0167-25G
G0167-500G
TCIAG0167-25GEA
29
EUR
TCIAG0167-25G
TCIAG0167-500G
Glycocyamine ≥97.0% (by titrimetric analysis)
Glycocyamine
Formula:
C₃H₇N₃O₂ MW: 117,11 g/mol |
Número MDL:
MFCD00004278 Núm. CAS: 352-97-6 EINECS: 206-529-5 Índice Merck: 14,04495 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 97.0 % |
Solubility in hot Water | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...