Glycyl-L-leucine ≥99.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
(S)-2-(2-Aminoacetamido)-4-methylpentanoic acid, H-Gly-Leu-OH
G0181-1G
G0181-5G
TCIAG0181-1GEA
61
EUR
TCIAG0181-1G
TCIAG0181-5G
Glycyl-L-leucine ≥99.0% (by titrimetric analysis)
Glycyl-L-leucine
Formula:
C₈H₁₆N₂O₃ MW: 188,23 g/mol |
Número MDL:
MFCD00008127 Núm. CAS: 869-19-2 EINECS: 212-785-9 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 99.0 % |
Specific rotation [a]20/D | -33.0 to -37.0 deg (C=2, H2O) |
Learn more
![](/m-es.vwr.com/es_ES/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...