Guaiacylglycerol-β-guaiacyl ether ≥97.0% (por GC)
Proveedor: TCI
Sinónimos:
1-(4-Hydroxy-3-methoxyphenyl)-2-(2-methoxyphenoxy)propane-1,3-diol, 1-(4-Hydroxy-3-methoxyphenyl)-2-(2-methoxyphenoxy)-1,3-propanediol
G0233-1G
G0233-100MG
TCIAG0233-1GEA
732
EUR
TCIAG0233-1G
TCIAG0233-100MG
Guaiacylglycerol-β-guaiacyl ether ≥97.0% (por GC)
Guaiacylglycerol-beta-guaiacyl ether
Formula:
C₁₇H₂₀O₆ |
Número MDL:
MFCD00060144 Núm. CAS: 7382-59-4 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 99.0 to 104.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...