L-(+)-Gulonic acid-γ-lactone ≥98.0% (por GC)

Proveedor: TCI

Sinónimos: L-Gulonic acid-1,4-lactone, (3S,4R,5R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxydihydrofuran-2(3H)-one, L-Gulonolactone

G0235-25G G0235-5G
TCIAG0235-25GEA 191 EUR
TCIAG0235-25G TCIAG0235-5G
L-(+)-Gulonic acid-γ-lactone ≥98.0% (por GC)
L-(+)-Gulonic acid-gamma-lactone
Formula: C₆H₁₀O₆
MW: 178,14 g/mol
Número MDL: MFCD00064331
Núm. CAS: 1128-23-0

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 184.0 to 188.0 deg-C
Specific rotation [a]20/D +55.0 to +57.0 deg (C=2, H2O)
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR