(R)-Glycidyl trityl ether ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
Sinónimos:
(R)-2-(Triphenylmethoxymethyl)oxirane, (R)-(+)-Glycidyl trityl ether, (R)-Glycidyl Triphenylmethyl Ether
G0284-25G
G0284-5G
TCIAG0284-25GEA
214
EUR
TCIAG0284-25G
TCIAG0284-5G
(R)-Glycidyl trityl ether ≥98.0% (por GC)
(R)-Glycidyl trityl ether
Formula:
C₂₂H₂₀O₂ MW: 316,40 g/mol |
Número MDL:
MFCD00273368 Núm. CAS: 65291-30-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Optical purity (LC) | min. 98.0 ee% |
Melting point | 99.0 to 103.0 deg-C |
Specific rotation [a]20/D | +10.0 to +12.0 deg (C=1, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...