(S)-Oxiran-2-ylmethyl-3-nitrobenzenesulphonate ≥98.0% (por GC)

Proveedor: TCI
Peligro

Sinónimos: (S)-(+)-Glycidyl nosylate, (S)-Oxiran-2-ylmethyl 3-nitrobenzenesulphonate, Glycidyl (S)-(+)-3-nitrobenzenesulphonate

G0287-25G G0287-5G
TCIAG0287-25GEA 134 EUR
TCIAG0287-25G TCIAG0287-5G
(S)-Oxiran-2-ylmethyl-3-nitrobenzenesulphonate ≥98.0% (por GC)
(S)-Oxiran-2-ylmethyl-3-nitrobenzenesulphonate
Formula: C₉H₉NO₆S
MW: 259,23 g/mol
Número MDL: MFCD00064582
Núm. CAS: 115314-14-2
UN: 1325
ADR: 4.1,II

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%
Melting point 63.0 to 66.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR