4-Glycidyloxycarbazole ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI
Atención

Sinónimos: 4-(2,3-Epoxypropoxy)carbazole, Carvedilol Related Compound D

G0296-25G G0296-5G
TCIAG0296-25GEA 136 EUR
TCIAG0296-25G TCIAG0296-5G
4-Glycidyloxycarbazole ≥98.0% (by HPLC, total nitrogen)
4-Glycidyloxycarbazole
Formula: C₁₅H₁₃NO₂
MW: 239,27 g/mol
Número MDL: MFCD03411884
Núm. CAS: 51997-51-4

Order Now

Resultados test especificaciones

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Solubility in THF almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR