Gal[2346Ac]β(1-3)GlcNPhth[46Bzd]-β-MP ≥97.0% (by HPLC)
Proveedor: TCI
Sinónimos:
4-Methoxyphenyl 3-O-(2,3,4,6-Tetra-O-acetyl-β-D-galactopyranosyl)-4,6-O-benzylidene-2-deoxy-2-phthalimido-β-D-glucopyranoside
G0311-1G
G0311-5G
TCIAG0311-1GEA
107
EUR
TCIAG0311-1G
TCIAG0311-5G
Gal[2346Ac]β(1-3)GlcNPhth[46Bzd]-β-MP ≥97.0% (by HPLC)
Gal[2346Ac]beta(1-3)GlcNPhth[46Bzd]-beta-MP
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Specific rotation [a]20/D | +9.0 to +13.0 deg (C=1, CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...