Glibenclamide ≥98.5% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: 5-Chloro-N-(4-(N-(cyclohexylcarbamoyl)sulphamoyl)phenethyl)-2-methoxybenzamide, 5-Chloro-N-[4-(3-cyclohexylureidosulphonyl)phenethyl]-2-methoxybenzamide, Glyburide

G0382-25G G0382-5G
TCIAG0382-25GEA 66 EUR
TCIAG0382-25G TCIAG0382-5G
Glibenclamide ≥98.5% (by HPLC, titration analysis)
Glibenclamide
Formula: C₂₃H₂₈ClN₃O₅S
MW: 494,00 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00056625
Núm. CAS: 10238-21-8
EINECS: 233-570-6
Índice Merck: 14,04478

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.5 area%
Purity (Neutralization titration) min. 98.5 %
Melting point 172.0 to 176.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR