Guanidine hydrobromide ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: Guanidine Hydrobromide

G0449-1G G0449-5G
TCIAG0449-1GEA 41 EUR
TCIAG0449-1G TCIAG0449-5G
Guanidine hydrobromide ≥98.0% (by titrimetric analysis)
Guanidine hydrobromide
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c49d7973f31bd721f9a1730a4682fce325e4269&vid=7c49d7973f31bd721f9a1730a4682fce325e4269&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 189.0 to 193.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR