Guanabenz Acetate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Peligro

Sinónimos: 2-(2,6-Dichlorobenzylidene)hydrazine-1-carboximidamide acetate

G0456-200MG G0456-1G
TCIAG0456-200MGEA 58 EUR
TCIAG0456-200MG TCIAG0456-1G
Guanabenz Acetate ≥98.0% (by HPLC, titration analysis)
Guanabenz Acetate
Formula: C₈H₈Cl₂N₄·C₂H₄O₂
MW: 291.13 g/mol
Número MDL: MFCD00153801
Núm. CAS: 23256-50-0
EINECS: 245-534-7
UN: 2811
ADR: 6.1 III
Índice Merck: 14,04558
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38861d8c1dcff9d39a22e389fcc75b4d098491c2&vid=38861d8c1dcff9d39a22e389fcc75b4d098491c2&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 188.0 to 192.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR