Graphene, nanoplatelets, Length: 5 µm, Ø 2-10 nm
Proveedor: TCI
|
Atención
|
G0499-1G
G0499-5G
TCIAG0499-1GEA
103
EUR
TCIAG0499-1G
TCIAG0499-5G
Graphene, nanoplatelets, Length: 5 µm, Ø 2-10 nm
Graphene
Número MDL:
MFCD00144065 Núm. CAS: 1034343-98-0 |
Resultados test especificaciones
Appearance | Light gray to Brown to Black powder to crystal |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...