Gadobutrol monohydrate ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: Gadolinium(III) 2,2',2''-[10-(1,3,4-Trihydroxybutan-2-yl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl]triacetate Monohydrate, Gadobutrol hydrate, Gadolinium(III) 2,2',2''-[10-(1,3,4-Trihydroxybutan-2-yl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl]triacetate

G0503-100MG G0503-25MG
TCIAG0503-100MGEA 245 EUR
TCIAG0503-100MG TCIAG0503-25MG
Gadobutrol monohydrate ≥98.0% (by titrimetric analysis)
Gadobutrol monohydrate
Formula: C₁₈H₃₁GdN₄O₉·H₂O
MW: 604,72 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD29917846
Núm. CAS: 198637-52-4

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 % (calcd.on anh.substance)
Elemental analysis (Nitrogen) 8.70 to 9.20 %
Water 2.6 to 3.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR