GBR 12909 dihydrochloride ≥98.0% (by titrimetric analysis)

Proveedor: TCI

G0541-250MG G0541-50MG
TCIAG0541-250MGEA 523 EUR
TCIAG0541-250MG TCIAG0541-50MG
GBR 12909 dihydrochloride ≥98.0% (by titrimetric analysis)
GBR 12909 dihydrochloride
Formula: C₂₈H₃₂F₂N₂O·2HCl
MW: 523,49 g/mol
Número MDL: MFCD00055193
Núm. CAS: 67469-78-7

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Melting point 219.0 to 223.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR