Homatropine hydrochloride ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

H0156-1G H0156-25G
TCIAH0156-1GEA 16 EUR
TCIAH0156-1G TCIAH0156-25G
Homatropine hydrochloride ≥98.0% (by titrimetric analysis)
Homatropine hydrochloride
Formula: C₁₆H₂₁NO₃
MW: 311,81 g/mol
Número MDL: MFCD00050613
Núm. CAS: 637-21-8
EINECS: 211-280-0
UN: 1544
ADR: 6.1,III
Índice Merck: 14,04730

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point gt. 217.0 to 223.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR