Benzenesulphohydroxamic acid ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
H0203-25G
H0203-5G
TCIAH0203-25GEA
187
EUR
TCIAH0203-25G
TCIAH0203-5G
Benzenesulphohydroxamic acid ≥98.0% (by HPLC, titration analysis)
Benzenesulphohydroxamic acid
Formula:
C₆H₇NO₃S |
Número MDL:
MFCD00025013 Núm. CAS: 599-71-3 EINECS: 209-973-8 Índice Merck: 14,07426 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...