Sodium-4-hydroxybenzoate ≥99.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: 4-Hydroxybenzoate sodium, Sodium 4-hydroxybenzoate, p-Hydroxybenzoic acid Monosodium Salt, 4-Hydroxybenzoic Acid Sodium Salt

H0221-25G H0221-500G
TCIAH0221-25GEA 24 EUR
TCIAH0221-25G TCIAH0221-500G
Sodium-4-hydroxybenzoate ≥99.0% (by titrimetric analysis)
Sodium-4-hydroxybenzoate
Formula: C₇H₆O₃
MW: 160.10 g/mol
Número MDL: MFCD00016530
Núm. CAS: 114-63-6
EINECS: 204-051-1

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Nonaqueous Titration) min. 99.0 %
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR