N-(2-Hydroxyethyl)ethylenediamine-N,N',N'-triacetic acid ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
2,2-((2-((Carboxymethyl)(2-hydroxyethyl)amino)ethyl)azanediyl)diacetic acid, N-(2-Hydroxyethyl)ethylenediaminetriacetic acid, EDTA-OH
H0243-25G
H0243-500G
H0243-100G
TCIAH0243-25GEA
19
EUR
TCIAH0243-25G
TCIAH0243-500G
TCIAH0243-100G
N-(2-Hydroxyethyl)ethylenediamine-N,N',N'-triacetic acid ≥98.0% (by titrimetric analysis)
N-(2-Hydroxyethyl)ethylenediamine-N,N',N'-triacetic acid
Formula:
C₁₀H₁₈N₂O₇ |
Número MDL:
MFCD00004294 Núm. CAS: 150-39-0 EINECS: 205-759-3 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Chelometric Titration) | min. 98.0 % |
Solubility in hot Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...