N-(3-Hydroxypropyl)ethylenediamine ≥97.0% (by GC, titration analysis)
Proveedor: TCI
|
Peligro
|
H0391-25G
H0391-5G
TCIAH0391-25GEA
588
EUR
TCIAH0391-25G
TCIAH0391-5G
N-(3-Hydroxypropyl)ethylenediamine ≥97.0% (by GC, titration analysis)
N-(3-Hydroxypropyl)ethylenediamine
Formula:
C₅H₁₄N₂O Pto. Ebullición: 149°C |
Número MDL:
MFCD00059848 Núm. CAS: 56344-32-2 UN: 3259 ADR: 8,III |
Resultados test especificaciones
Appearance | White or Colorless to Yellow powder to lump to clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 46.0 to 51.0 deg-C |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...