2-Hexadecenoic acid ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
H0428-1G
H0428-5G
TCIAH0428-1GEA
131
EUR
TCIAH0428-1G
TCIAH0428-5G
2-Hexadecenoic acid ≥98.0% (by titrimetric analysis)
2-Hexadecenoic acid
Formula:
C₁₆H₃₀O₂ |
Número MDL:
MFCD00045907 Núm. CAS: 629-56-1 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 46.0 to 51.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...