Homatropine methyl bromide

Proveedor: TCI
Atención

Sinónimos: N-Methylhomatropinium Bromide

H0448-25G H0448-5G
TCIAH0448-25GEA 260 EUR
TCIAH0448-25G TCIAH0448-5G
Homatropine methyl bromide
Homatropine methyl bromide
Formula: C₁₇H₂₄BrNO₃
MW: 370,29 g/mol
Número MDL: MFCD00050292
Núm. CAS: 80-49-9
EINECS: 201-284-0
UN: 1544
ADR: 6.1,III
Índice Merck: 14,04730

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 193.0 to 198.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR