2-Hydroxypyrimidine hydrochloride ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: Pyrimidin-2-ol hydrochloride, 2-Pyrimidinol Hydrochloride, 2(1H)-pyrimidinone monohydrochloride

H0480-25G H0480-250G
TCIAH0480-25GEA 23 EUR
TCIAH0480-25G TCIAH0480-250G
2-Hydroxypyrimidine hydrochloride ≥98.0% (by HPLC, titration analysis)
2-Hydroxypyrimidine hydrochloride
Formula: C₄H₄N₂O
MW: 132,55 g/mol
Número MDL: MFCD00012781
Núm. CAS: 38353-09-2
EINECS: 253-897-8

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR