Methyl Hyodeoxycholate ≥96.0% (por GC)

Proveedor: TCI

Sinónimos: (R)-Methyl 4-((3R,5R,6S,8S,9S,10R,13R,14S,17R)-3,6-dihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoate, Hyodeoxycholic acid methyl ester

H0529-25G H0529-5G
TCIAH0529-25GEA 319 EUR
TCIAH0529-25G TCIAH0529-5G
Methyl Hyodeoxycholate ≥96.0% (por GC)
Methyl Hyodeoxycholate
Formula: C₂₅H₄₂O₄
MW: 406,61 g/mol
Número MDL: MFCD00059509
Núm. CAS: 2868-48-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Specific rotation [a]20/D +7.0 to +9.0 deg (C=1, EtOH)
Residual Toluene max. 1.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR