Sodium-3-(4-hydroxy-3-methoxyphenyl)lactate ≥99.0% (by HPLC)

Proveedor: TCI

Sinónimos: 3-(4-Hydroxy-3-methoxyphenyl)lactic Acid Sodium Salt, Sodium 2-Hydroxy-3-(4-hydroxy-3-methoxyphenyl)propionate, 2-Hydroxy-3-(4-hydroxy-3-methoxyphenyl)propionic Acid Sodium Salt

H0538-1G H0538-100MG
TCIAH0538-1GEA 516 EUR
TCIAH0538-1G TCIAH0538-100MG
Sodium-3-(4-hydroxy-3-methoxyphenyl)lactate ≥99.0% (by HPLC)
Sodium-3-(4-hydroxy-3-methoxyphenyl)lactate
Formula: C₁₀H₁₂O₅
MW: 234,18 g/mol
Número MDL: MFCD00056719
Núm. CAS: 77305-02-3

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Ion exchange titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR