DL-4-Hydroxyphenyllactic acid ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

H0542-1G H0542-100MG
TCIAH0542-1GEA 409 EUR
TCIAH0542-1G TCIAH0542-100MG
DL-4-Hydroxyphenyllactic acid ≥98.0% (by HPLC, titration analysis)
DL-4-Hydroxyphenyllactic acid
Formula: C₉H₁₀O₄
Número MDL: MFCD00010514
Núm. CAS: 306-23-0

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Water max. 6.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR